Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Paraformaldehyde 4%
SDP

Catalog No. 22023770
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-770 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-770 Supplier StatLab Supplier No. S23341GL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Poly Scientific Paraformaldehyde 4% in 0.1 M Phosphate Buffer pH 7.4 - a fixative often used for immunohistochemistry and electron microscopy

  • It can be used to preserve the structure and integrity of tissues during the perfusion process
  • It is also effective in fixing and preserving tissues for IHC, allowing for the detection of specific proteins and antigens
  • It can be used in IF to fix and preserve tissues for the detection of fluorescently labeled antibodies
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

Concentration 4%
For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Fixative
Quantity 1 gal.
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.