Learn More
StatLab Poly Scientific ParaWay Paraffin Remover Solution
SDP
Catalog No. 21010266
Change view
Click to view available options
Form:
Solution
Catalog No. Form
21-010-266 Solution
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-266 Supplier StatLab Supplier No. CPW4426CT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Convenient spray bottle.
  • Simply spray the solution onto work surfaces where paraffin routinely builds up and wipe it away
  • ParaWay is a must have at every histotechs embedding and cutting stations
  • Citrus scented
  • 100 mL spray bottle
Specifications
Product Type Paraffin Remover
Quantity 100 mL
Form Solution