Learn More
StatLab Poly Scientific PAS with Diastase Digestion Special Stain Kit

Description
- The diastase enzyme acts on glycogen, de-polymerizing it into smaller sugar units that are washed out of the section
- This method is particularly useful for diagnosing conditions such as glycogen storage disorders, certain types of cancers, and fungal infections
- The PAS stain provides a magenta coloration in the presence of glycogen, making it easier for pathologists to identify and differentiate between normal and abnormal tissue components
- A StatLab - Poly Scientific Special Stain Kit
Specifications
Specifications
| CAS | 67-56-1 |
| Color | Assorted |
| Form | Solution |
| Quantity | 16 oz. |
| Product Type | Stain Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.