Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Periodic Acid 1% Aqueous
SDP

Catalog No. 23640012
Change view
Click to view available options
Quantity:
8 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-012 8 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-012 Supplier StatLab Supplier No. S18618OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

Used in the Periodic Acid-Schiff (PAS) staining process for tissue component visualization.

  • Used in the Periodic Acid-Schiff (PAS) staining method, a fundamental histological technique for detecting and visualizing key tissue components.
  • The PAS staining process highlights structures such as glycogen, mucopolysaccharides, basement membranes, mucoproteins, and fungi, supporting detailed tissue analysis and diagnosis.
  • Provided as a StatLab Poly Scientific Special Stain Kit component, ensuring consistent quality, compatibility, and reliable staining performance in laboratory workflows.

Specifications

Quantity 8 oz.
Product Type Stain Kit Component
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.