Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Periodic Acid 1% Aqueous
SDP

Catalog No. 22021451
Change view
Click to view available options
Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
22-021-451 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-451 Supplier StatLab Supplier No. S186116OZ
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Used to oxidize glycols to aldehydes.

  • A crucial component in the Periodic Acid-Schiff (PAS) staining method
  • This method is used to detect and stain various polysaccharides in tissues, including glycogen, neutral mucous substances, basement membranes, and other polysaccharides
  • The solution oxidizes these compounds, releasing aldehyde groups that react with the Schiff reagent, resulting in a magenta staining visible under the microscope
  • Commonly used with the Alcian Blue PAS
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

For Use With (Equipment) Staining Equipment
Grade Chemical Solution
Product Type Periodic Acid
Quantity 16 oz.
Solution Type Oxidizer
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.