Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Phosphate Buffered Saline 0.1M pH 7.2
SDP

Catalog No. 22021463
Change view
Click to view available options
Grade:
Laboratory
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
22-021-463 Laboratory
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-463 Supplier StatLab Supplier No. S23241GL
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

For preserving tissues during surgical procedures and IHC staining procedures.

  • It serves as an important fixative for light and electron microscopy, ensuring that tissues remain intact for analysis
  • Also used in various staining procedures to enhance the visibility of cellular structures
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

For Use With (Equipment) Staining Equipment
Quantity 1 gal.
Solution Type Buffer
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.