Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab Poly Scientific Reagent Alcohol 90%

Description
- Specially denatured 100% alcohol with 10% parts of deionized water added to make this ready to use 90% graded alcohol
- Suitable for use in clinical laboratory protocols that reference 90% alcohol or 90% Ethanol
- Ideal for processing and staining protocols in Histology and Cytology laboratories
- Case of 4 gal.
Specifications
Specifications
| Product Type | Alcohol |
| Quantity | 4 gal. |
| Form | Solution |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?