Learn More
StatLab Poly Scientific Rhodanine Stock Solution

Description
- The Rhodanine stock solution is primarily used for detecting copper and copper-associated proteins (CAP) in tissue sections
- It acts as a bidentate chelating agent, effectively binding to copper deposits, allowing for their visualization in histological preparations
- This method is particularly useful in identifying excess copper in cells, which can be indicative of various pathological conditions
- A StatLab - Poly Scientific Special Stain Kit Component
Specifications
Specifications
| Chemical Name or Material | Rhodadine Stock Solution |
| Color | Reddish Brown |
| Quantity | 32 oz. |
| Grade | Histological |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.