Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Rhodanine Working Solution
SDP

Catalog No. 22023760
Change view
Click to view available options
Quantity:
32 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-760 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-760 Supplier StatLab Supplier No. S204432OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Poly Scientific Rhodanine Working Solution - used for differentiating collagen and muscle fibers in tissue sections

  • Used for differentiating collagen and muscle fibers in tissue sections, particularly in the study of pathologies such as heart infarction, liver cirrhosis, and kidney globular fibrosis
  • Also used for identifying copper deposits in tissues, which can indicate conditions like Wilson's Disease
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

Chemical Name or Material Rhodadine Working Solution
Grade Histological
Color White
Quantity 32 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.