Learn More
StatLab Poly Scientific Schiff Reagent
SDP
Catalog No. 22021465
Change view
Click to view available options
Product Type:
Periodic Acid Schiff
Catalog No. Product Type
22-021-465 Periodic Acid Schiff
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-465 Supplier StatLab Supplier No. S27216OZ
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Used in histology for the Periodic Acid-Schiff (PAS) reaction.
  • The PAS reaction is utilized to demonstrate polysaccharides, neutral mucosubstances, basement membranes, and fungal cell walls
  • A StatLab - Poly Scientific Special Stain Kit Component
Specifications
Quantity 16 oz.
Product Type Periodic Acid Schiff