Learn More
StatLab Poly Scientific Silver Nitrate Solution (Fontana)
SDP
Catalog No. 22033332
Change view
Click to view available options
Quantity:
4 oz.
Catalog No. Quantity
22-033-332 4 oz.
1 options
This item is not returnable. View return policy
Catalog No. 22-033-332 Supplier StatLab Supplier No. S181S4OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used with fontana masson silver method for argentaffin granule
  • Silver Nitrate solution is used with Fontana Masson Silver Method for Argentaffin Granules
  • Argentaffin substances are able to bind and reduce silver to the visible form
  • A StatLab - Poly Scientific Special Stain Kit Component
Specifications
Physical Form Solution
Quantity 4 oz.