Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Sodium Chloride 0.9% Aqueous
SDP

Catalog No. 22021460
Change view
Click to view available options
Physical Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Physical Form
22-021-460 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-460 Supplier StatLab Supplier No. S19165GL
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Used in Congo Red staining procedures for the demonstration of amyloid in tissue.

  • Used as an isotonic carrier medium for other stains and solutions
  • It's isotonic properties help preserve the structure of cells and tissue during the staining process, preventing the damage that can be caused by osmotic pressure
  • Used as a diluent for stains, for rinsing and washing, and for tissue fixation
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

Physical Form Solution
CAS 67-56-1, 67-63-0, 64-19-7, 64-17-5, 50-00-0
Color Colorless
Quantity 5 gal.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.