Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Sodium Chloride Saturated in 80% Reagent Alcohol
SDP

Catalog No. 22023753
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-753 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-753 Supplier StatLab Supplier No. S249816OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Poly Scientific Sodium Chloride Saturated in 80% Reagent Alcohol - Sodium Chloride Saturated in 80% Reagent Alcohol

  • Sodium chloride helps regulate the concentration of sodium and chloride ions in extracellular fluids, which is crucial for maintaining cell volume and structural integrity
  • It is integral to the sodium-potassium pump, which actively transports sodium ions out of the cell and potassium ions into the cell, consuming ATP in the process
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

Color Colorless
Quantity 16 oz.
Grade Histological
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.