Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Sodium Metabisulfite 1% Aqueous
SDP

Catalog No. 21402151
Change view
Click to view available options
Quantity:
237 mL
473 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-402-151 473 mL
21-402-152 237 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-402-151 Supplier StatLab Supplier No. S27816OZ
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Sodium metabisulfite, used for its ability to preserve tissues and prevent oxidation

  • Solution is particularly useful in procedures like Gomori methenamine staining, which is used for the detection of fungi in tissue sections
  • The 1% solution is created by dissolving 1 gram of sodium metabisulfite in 100 milliliters of water and is known for its acidic pH, making it suitable for specific staining applications

Specifications

Physical Form Solution
Color Colorless
CAS 7681-57-4
Quantity 473 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.