Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Southgate's Mucicarmine Stock Solution
SDP

Catalog No. 22033333
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-033-333 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-033-333 Supplier StatLab Supplier No. S216116OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used in Southgate's Mucicarmine Method for the qualitative demonstration of mucin or Cryptococcus

  • A component of the Mucicarmine Stain Kit, which is designed for the histological visualization of acid mucopolysaccharides in tissue sections
  • This stain is particularly useful in distinguishing mucin negative undifferentiated squamous cell lesions from mucin positive adenocarcinomas and staining the mucopolysaccharide capsule of Cryptococcus neoformans
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

Physical Form Solution
CAS 67-56-1, 64-17-5, 67-63-0, 7446-70-0, 21645-51-2, 1390-65-4
Color Colorless
Quantity 16 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.