Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Special Stain Kit Component, Ether Acetone 1:1
SDP

Catalog No. 23640209
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-209 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-209 Supplier StatLab Supplier No. S178P32OZ
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Ether Acetone 1:1

  • A StatLab - Poly Scientific Special Stain Kit Component
  • Used in histology lab applications for Brown and Brenn Stain for Bacteria
  • Also suitable for histology-related applications and is used for tissue processing and dehydration

Specifications

Physical Form Aqueous Solution
Color Red
Quantity 32 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.