Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Special Stain Kit Component, Fast Green Substitute For Light Green 0.2% Aqueous
SDP

Catalog No. 23640241
Change view
Click to view available options
Quantity:
8 oz.
16 oz.
32 oz.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-241 8 oz.
23-640-240 32 oz.
23-640-239 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-241 Supplier StatLab Supplier No. S2328OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Fast Green Substitute For Light Green 0.2% Aqueous

  • A StatLab - Poly Scientific Special Stain Kit Component
  • Used as a counterstain in GMS stains

Specifications

Quantity 8 oz.
Product Type Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.