Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Special Stain Kit Component, Ferric Ammonium Sulfate 0.2% Aqueous
SDP

Catalog No. 23640210
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-210 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-210 Supplier StatLab Supplier No. S1798OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Ferric Ammonium Sulfate 0.2% Aqueous

  • A StatLab - Poly Scientific Special Stain Kit Component
  • Used as a sensitizer in silver staining, and for reticulum stains to demonstrate reticulum fibers
  • Also used in hematoxylin and eosin (H and E) staining to obtain a detailed view of tissue

Specifications

Physical Form Aqueous Solution
Color Red
Quantity 8 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.