Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Special Stain Kit Component, Lugol's Iodine Working Solution
SDP

Catalog No. 23640244
Change view
Click to view available options
Quantity:
16 oz.
32 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-244 16 oz.
23-640-245 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-244 Supplier StatLab Supplier No. S234A16OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Lugol's Iodine Working Solution

  • A StatLab - Poly Scientific Special Stain Kit Component
  • Lugol's Iodine Working Solution acts as a mordant and be used inmost Elastic and Gram stain procedures

Specifications

Physical Form Aqueous Solution
Color Brown
Quantity 16 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.