Learn More
StatLab Poly Scientific Special Stain Kit Component, Mayers Modified Hematoxylin

Description
- A StatLab - Poly Scientific Special Stain Kit Component
- Can be used in H and E staining procedures and in many Special Stains procedures as a counterstain
Specifications
Specifications
| Physical Form | Aqueous Solution |
| Color | Purple/Violet |
| Quantity | 16 oz. |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.