Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Special Stain Kit Component, Mayers Modified Hematoxylin
SDP

Catalog No. 23640234
Change view
Click to view available options
Quantity:
16 oz.
32 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-234 32 oz.
23-640-233 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-234 Supplier StatLab Supplier No. S21632OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Mayers Modified Hematoxylin

  • A StatLab - Poly Scientific Special Stain Kit Component
  • Can be used in H and E staining procedures and in many Special Stains procedures as a counterstain

Specifications

Physical Form Aqueous Solution
Color Purple/Violet
Quantity 32 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.