Learn More
StatLab Poly Scientific Special Stain Kit Component, Methenamine Silver Nitrate Stock Solution Set (A and B)

Description
- A StatLab - Poly Scientific Special Stain Kit Component
- Use with Gomori's Methenamine Silver Nitrate for Fungi and Pneumocystis carinii. Contains both Stock Solution A and B
Specifications
Specifications
| Physical Form | Aqueous Solution |
| Color | Clear Colorless |
| Quantity | 16 oz. |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.