Learn More
StatLab Poly Scientific Special Stain Kit Component, Methylene Blue Working Solution
SDP
Catalog No. 23640221
Change view
Click to view available options
Quantity:
8 oz.
16 oz.
32 oz.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-221 8 oz.
23-640-219 16 oz.
23-640-220 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-221 Supplier StatLab Supplier No. S188B8OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Methylene Blue Working Solution
  • A StatLab - Poly Scientific Special Stain Kit Component
  • Can be used as a counterstain in AFB stain procedures
Specifications
Quantity 8 oz.
Product Type Stain Kit