Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Special Stain Kit Component, Poly Mount (Xylene Based)
SDP

Catalog No. 23640232
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-232 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-232 Supplier StatLab Supplier No. S215316OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Poly Mount (Xylene Based), an acrylic medium used for histology and cytology slides

  • A StatLab - Poly Scientific Special Stain Kit Component
  • It provides superior optical clarity and is recommended for automated coverslipping instruments that use glass coverslips
  • It is non-fluorescing and non-permanent, making it suitable for mounting sections directly from aqueous solutions

Specifications

Physical Form Liquid
Color Clear Colorless
Quantity 16 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.