Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Special Stain Kit Component, Potassium Ferrocyanide 2% Aqueous
SDP

Catalog No. 23640216
Change view
Click to view available options
Quantity:
16 oz.
32 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-216 16 oz.
23-640-217 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-216 Supplier StatLab Supplier No. S187216OZ
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Potassium Ferrocyanide 2% Aqueous

  • A StatLab - Poly Scientific Special Stain Kit Component
  • Used for Prussian blue staining
  • It reacts with ferric iron to produce an insoluble blue pigment called Turnbull blue
  • The solution is used to highlight the presence of ferric iron in tissue sections

Specifications

Physical Form Aqueous Solution
Color Yellow
Quantity 16 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.