Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Special Stain Kit Component, Potassium Hydroxide 20% Aqueous
SDP

Catalog No. 23640218
Change view
Click to view available options
Color:
Clear Colorless
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-218 Clear Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-218 Supplier StatLab Supplier No. S18761GL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Potassium Hydroxide 20% Aqueous

  • A StatLab - Poly Scientific Special Stain Kit Component
  • Used for tissue and cell staining
  • It is employed in the preparation of biological samples for microscopic analysis and in the identification and differentiation of cellular structures

Specifications

Physical Form Aqueous Solution
Color Clear Colorless
Quantity 1 gal.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.