Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Special Stain Kit Component, Sodium Bisulfite 1% Aqueous
SDP

Catalog No. 23640222
Change view
Click to view available options
Color:
Clear Colorless
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-222 Clear Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-222 Supplier StatLab Supplier No. S189316OZ
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Sodium Bisulfite 1% Aqueous

  • A StatLab - Poly Scientific Special Stain Kit Component
  • Used in GMS staining procedures for the qualitative demonstration of fungi and Pneumocystis carinii

Specifications

Physical Form Aqueous Solution
Color Clear Colorless
Quantity 16 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.