Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Special Stain Kit Component, Sodium Thiosulfate 5% Aqueous
SDP

Catalog No. 23640223
Change view
Click to view available options
Color:
Clear Colorless
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-223 Clear Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-223 Supplier StatLab Supplier No. S18958OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Sodium Thiosulfate 5% Aqueous

  • A StatLab - Poly Scientific Special Stain Kit Component
  • Used in silver staining procedures to remove excess silver

Specifications

Physical Form Aqueous Solution
Color Clear Colorless
Quantity 8 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.