Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Stain Kit, Brown Hopps Method For Gram +/- Bacteria Stain Kit
SDP

Catalog No. 23640179
Change view
Click to view available options
Color:
Colorless
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-179 Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-179 Supplier StatLab Supplier No. K0058OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Brown Hopps Method For Gram +/- Bacteria stain kit

  • For demonstrating gram-negative and gram-positive in tissue
  • Both bacteria's, positive and negative, cell wall is composed of peptidoglycan, and both will take up the crystal violet

Specifications

Physical Form Aqueous Solution
Color Colorless
Quantity 8 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.