Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Weigerts Hematoxylin Solution A
SDP

Catalog No. 22023784
Change view
Click to view available options
Quantity:
8 oz.
32 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-784 8 oz.
22-023-779 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-784 Supplier StatLab Supplier No. S216BA8OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Poly Scientific Weigerts Hematoxylin Solution A - an iron hematoxylin that stains nuclei black or dark purple

  • Weigert's Hematoxylin Solution A is an iron hematoxylin that stains nuclei black or dark purple
  • It is often used in trichrome stains and is part of the van Gieson's trichrome staining procedure
  • The solution is mixed in equal parts with Weigert's Hematoxylin Solution B to form the working solution
  • The final color of the nuclei ranges from black to dark purple, depending on the staining technique and the tissue being examined
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

Physical Form Aqueous Solution
Color Dark Violet
Quantity 8 oz.
Grade Histological
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.