Learn More
StatLab Poly Scientific Xylene Peanut Oil 2:1 Fite's Method
SDP
Catalog No. 21010352
Change view
Click to view available options
Form:
Solution
Catalog No. Form
21-010-352 Solution
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-352 Supplier StatLab Supplier No. S1912A32OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
A histological technique used to stain acid-fast organisms, particularly in the diagnosis of leprosy.
  • This method involves the use of a solution of xylene and peanut oil to deparaffinize tissue while preserving the acid-fastness of the bacteria
  • The technique is crucial for identifying Mycobacterium leprae, the causative agent of leprosy, as these bacteria have cell walls containing mycolic acids that are resistant to decolorization with acid-alcohol
  • The peanut oil coats the bacteria, protecting the waxy coat of the cell walls and ensuring the acid-fastness is maintained during the staining process
  • A StatLab - Poly Scientific Special Stain Kit Component
Specifications
Product Type Xylene
Quantity 32 oz.
Form Solution