Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Ziehl-Neelsen Method
SDP

Catalog No. 22033329
Change view
Click to view available options
Color:
Colorless
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-033-329 Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-033-329 Supplier StatLab Supplier No. K0628OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy

For acid fast bacteria special stain kit - intended for the qualitative demonstration of acid fast organisms

  • The Ziehl-Neelsen method is a crucial technique in histology analysis for identifying acid-fast bacteria (AFB), particularly those belonging to the Mycobacterium genus, which are the causative agents of tuberculosis (TB)
  • This method is essential for diagnosing TB and other diseases caused by atypical mycobacteria, such as leprosy and Mycobacterium avium-intracellular infection
  • Kit includes Carbol Fuchsin Ziehl Neelsen, Acid Alcohol 1%, and Methylene Blue Working solution
  • A StatLab - Poly Scientific Special Stain Kit

Specifications

Physical Form Solution
CAS 64-17-5, 67-63-0, 7647-01-0, 67-56-1
Color Colorless
Quantity 4 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.