Learn More
MSC Vikan UST Hand Brush, 6.9 in, Stiff, Purple, 1 ea
Catalog No. 502816653
Encompass
The Vikan UST Hand Brush with ULTRA SAFE TECHNOLOGY (UST) offers optimal cleaning for high-risk food-preparation surfaces, conveyors, and machinery. Its unique bristle system minimizes contamination and loss, while the ergonomic block reduces wrist strain.
  • Ideal for high-risk food manufacturing, retail, and service cleaning.
  • Stiff, thick bristles effectively scrub and dislodge stubborn soils.
  • Individual bristle molding ensures unmatched retention, minimizing contamination.
  • Optimized bristle pattern and hygienic design reduce cross-contamination.
  • Angled bristles reach corners and crevices; ergonomic design reduces strain.
  • Color-coded for hygienic zoning and 5S programs.
  • Easy to clean and maintain; drop-shaped hanging hole aids storage.
Catalog No. 50-281-6653 Supplier MSC Supplier No. 05083886
The Vikan UST Hand Brush with ULTRA SAFE TECHNOLOGY (UST) offers optimal cleaning for high-risk food-preparation surfaces, conveyors, and machinery. Its unique bristle system minimizes contamination and loss, while the ergonomic block reduces wrist strain.
  • Ideal for high-risk food manufacturing, retail, and service cleaning.
  • Stiff, thick bristles effectively scrub and dislodge stubborn soils.
  • Individual bristle molding ensures unmatched retention, minimizing contamination.
  • Optimized bristle pattern and hygienic design reduce cross-contamination.
  • Angled bristles reach corners and crevices; ergonomic design reduces strain.
  • Color-coded for hygienic zoning and 5S programs.
  • Easy to clean and maintain; drop-shaped hanging hole aids storage.