Learn More
MSC Vikan UST Hand Brush, 6.9 in, Stiff, Orange, 1 ea
Catalog No. 502818120
Encompass
This ULTRA SAFE TECHNOLOGY (UST) Hand Brush is designed for optimal cleaning of conveyor belts, production lines, machinery, and food-preparation surfaces in high-risk areas.
  • Angled Filament Security Units for effective cleaning
  • Unique bristle system minimizes contamination and bristle loss
  • Ergonomically designed brush block prevents slipping and reduces wrist strain
  • Effective surface contact allows strong pressure
  • Angled bristles are effective for cleaning corners, junctions, and hard-to-reach areas
Catalog No. 50-281-8120 Supplier MSC Supplier No. 05083928
This ULTRA SAFE TECHNOLOGY (UST) Hand Brush is designed for optimal cleaning of conveyor belts, production lines, machinery, and food-preparation surfaces in high-risk areas.
  • Angled Filament Security Units for effective cleaning
  • Unique bristle system minimizes contamination and bristle loss
  • Ergonomically designed brush block prevents slipping and reduces wrist strain
  • Effective surface contact allows strong pressure
  • Angled bristles are effective for cleaning corners, junctions, and hard-to-reach areas