Learn More
MSC Vikan Hand Brush, Stiff, White, 176 mm, 1 ea
Catalog No. 502815923
Encompass
This UST hand brush is designed for optimal cleaning in high-risk areas such as conveyor belts, production lines, machinery, and food preparation surfaces. Its unique bristle system minimizes contamination risk and bristle loss, ensuring effective cleaning.
  • Ergonomically designed brush block reduces wrist strain and prevents slipping
  • Angled bristles for effective cleaning in corners and hard-to-reach areas
  • Provides effective surface contact to apply hard pressure
  • Chemical resistant construction
  • FDA compliant raw materials
  • Halal and Kosher compliant
  • Designed for high-risk food production facilities
  • Minimizes risk of cross-contamination
Catalog No. 50-281-5923 Supplier MSC Supplier No. 05086038
This UST hand brush is designed for optimal cleaning in high-risk areas such as conveyor belts, production lines, machinery, and food preparation surfaces. Its unique bristle system minimizes contamination risk and bristle loss, ensuring effective cleaning.
  • Ergonomically designed brush block reduces wrist strain and prevents slipping
  • Angled bristles for effective cleaning in corners and hard-to-reach areas
  • Provides effective surface contact to apply hard pressure
  • Chemical resistant construction
  • FDA compliant raw materials
  • Halal and Kosher compliant
  • Designed for high-risk food production facilities
  • Minimizes risk of cross-contamination