Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(sodium 4-styrenesulfonate)

Catalog No. 243051100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-305-1100G 100 g
24-305-15G 5 g
24-305-1500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 24-305-1100G Supplier Sigma Aldrich Supplier No. 243051100G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(sodium 4-styrenesulfonate)

Specifications

CAS 25704-18-1
Melting Point 460°C (dec.)
Density 0.801 g/mL (at 25°C)
Linear Formula (C8H7NaO3S)n
MDL Number MFCD00084449
Quantity 100 g
Synonym PSS; Poly(4-styrenesulfonic acid) sodium salt
Molecular Weight (g/mol) 70000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.