Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(sodium 4-styrenesulfonate) solution

Catalog No. 561967500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-196-7500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-196-7500G Supplier Sigma Aldrich Supplier No. 561967500G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly (Sodium 4-Styrenesulfonate) (PSS), a strong anionic polyelectrolyte, possesses a number of negative charges along its backbone chain. The ionic charges, in such electrolytes, are largely independent of the pH condition. PSS deposition exhibits layer by layer assembly. Alternate adsorption of polycations and polyanions by electrostatic attraction forms polyelectrolyte multilayers (PEMs). PSS-graphite oxide shows high performance of electric double layer capacitance compared to pristine graphite oxide on account of the layer by layer assembly. PSS coating on nanoparticles inhibit formation of agglomeration.

Specifications

CAS 25704-18-1
Density 1.1471 g/mL (at 25°C)
Linear Formula (C8H7NaO3S)n
Refractive Index n20/D ~1.3961
Quantity 500 g
MDL Number MFCD00084449
Synonym PSS; Poly(4-styrenesulfonic acid) sodium salt
Molecular Weight (g/mol) 200000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.