Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(sodium 4-styrenesulfonate) solution

Catalog No. 527483100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-748-3100ML 100 mL
52-748-31L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52-748-3100ML Supplier Sigma Aldrich Supplier No. 527483100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Effective antistatic and specialty dispersant.

Specifications

CAS 25704-18-1
Density 1.158 g/mL (at 25°C)
Boiling Point 100°C
Refractive Index n20/D 1.395
Linear Formula (C8H7NaO3S)n
Quantity 100 mL
MDL Number MFCD00084449
Synonym PSS; Poly(4-styrenesulfonic acid) sodium salt
Molecular Weight (g/mol) 70000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.