Learn More
MSC PIP Cut, Puncture and Abrasion-Resistant Gloves, Cut A6, Puncture 3, Abrasion 4, Size Medium, 1 Pair
Catalog No. 5000508290
Encompass
These mechanic's style gloves are engineered to provide high-level hand protection in demanding environments requiring cut, puncture, and abrasion resistance. Constructed with a polyester base and featuring a durable thermoplastic elastomer back, these gloves offer a balance of safety and dexterity for industrial applications.
  • ANSI/ISEA cut resistance level A6
  • ANSI/ISEA abrasion resistance level 4
  • ANSI/ISEA puncture resistance level 3
  • Polyurethane grip surface on palm
  • Slip-on cuff style for ease of use
  • Size medium (8)
  • Blue and black color scheme
Catalog No. 50-005-08290 Supplier MSC Supplier No. 15003858
Add to Cart
Add to Cart
These mechanic's style gloves are engineered to provide high-level hand protection in demanding environments requiring cut, puncture, and abrasion resistance. Constructed with a polyester base and featuring a durable thermoplastic elastomer back, these gloves offer a balance of safety and dexterity for industrial applications.
  • ANSI/ISEA cut resistance level A6
  • ANSI/ISEA abrasion resistance level 4
  • ANSI/ISEA puncture resistance level 3
  • Polyurethane grip surface on palm
  • Slip-on cuff style for ease of use
  • Size medium (8)
  • Blue and black color scheme