Learn More
MSC Vikan Bucket, 6 Litre, Polypropylene, Stainless Steel, Purple
Catalog No. 502839469
Encompass
This durable 6-litre bucket is suitable for cleaning or food storage in industrial and commercial settings. Constructed from heavy-duty polypropylene and featuring a stainless steel handle, it includes a wide, drip-free spout, a bottom hand grip, and internal measurements for accurate use. Its design helps prevent spillage, and it is FDA compliant.
  • Durable 6-litre capacity
  • Wide, drip-free spout
  • Sturdy stainless steel handle
  • Bottom hand grip raised from the base
  • Calibrated with gallon and liter measurements for accurate use
  • Flat side helps prevent spillage
  • Suitable for cleaning or food storage
  • FDA compliant
  • Dishwasher safe
  • Stackable
  • Drainage hole prevents moisture accumulation when stored
  • Purple color for color-coded systems
Catalog No. 50-283-9469 Supplier MSC Supplier No. 47171921
This durable 6-litre bucket is suitable for cleaning or food storage in industrial and commercial settings. Constructed from heavy-duty polypropylene and featuring a stainless steel handle, it includes a wide, drip-free spout, a bottom hand grip, and internal measurements for accurate use. Its design helps prevent spillage, and it is FDA compliant.
  • Durable 6-litre capacity
  • Wide, drip-free spout
  • Sturdy stainless steel handle
  • Bottom hand grip raised from the base
  • Calibrated with gallon and liter measurements for accurate use
  • Flat side helps prevent spillage
  • Suitable for cleaning or food storage
  • FDA compliant
  • Dishwasher safe
  • Stackable
  • Drainage hole prevents moisture accumulation when stored
  • Purple color for color-coded systems