Learn More
MSC Simpson Hose, 3/8 in, 50 ft, 4500 PSI, 1 ea
Catalog No. 502869083
Encompass
This pressure washer hose is designed for cold water applications up to 140°F. It features dual-braided steel over polyester construction for strength and a translucent polyurethane outer jacket for flexibility and abrasion resistance.
  • Easy quick connect fittings (3/8 in male plug x 3/8 in female socket)
  • Dual-braided steel over polyester construction for durability
  • Flexible, non-marring, and abrasion-resistant polyurethane outer jacket
  • Compatible with cold water pressure washers up to 4500 PSI
  • Includes 4-inch bend protectors for extended accessory life
  • Wipes clean easily
Catalog No. 50-286-9083 Supplier MSC Supplier No. 32660193
This pressure washer hose is designed for cold water applications up to 140°F. It features dual-braided steel over polyester construction for strength and a translucent polyurethane outer jacket for flexibility and abrasion resistance.
  • Easy quick connect fittings (3/8 in male plug x 3/8 in female socket)
  • Dual-braided steel over polyester construction for durability
  • Flexible, non-marring, and abrasion-resistant polyurethane outer jacket
  • Compatible with cold water pressure washers up to 4500 PSI
  • Includes 4-inch bend protectors for extended accessory life
  • Wipes clean easily