Learn More
USA Scientific Inc 81-place polycarbonate box, assorted colors
Catalog No. NC9457957
Encompass
Polycarbonate 81-place box, assorted color bases with clear lid, 5.2 in L x 5.2 in W x 2.1 in H, 5/pack
Catalog No. NC9457957 Supplier USA Scientific Inc Supplier No. 23813508
Polycarbonate 81-place box, assorted color bases with clear lid, 5.2 in L x 5.2 in W x 2.1 in H, 5/pack