Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Poly(styrene)-block-poly(acrylic acid) (PS:PAA 30000:2000) | 90.0% | Polystyrene block: 30000, Polyacrylic acid block: 2000 | 25 MG
SDP

Catalog No. 5000366047
Encompass_Preferred

Poly(styrene)-block-poly(acrylic acid) (PS:PAA 30000:2000) is a diblock copolymer primarily utilized in the preparation of polymer micelles, vesicles, and other encapsulation applications. This product is intended for research use only.

  • Diblock copolymer
  • Used to prepare polymer micelles or vesicles
  • Suitable for encapsulation applications
  • Appearance: solid
  • Color: white to off-white
  • Purity: 90.0%

Catalog No. 50-003-66047 Supplier Medchemexpress LLC Supplier No. 25MGHY172378
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Poly(styrene)-block-poly(acrylic acid) (PS:PAA 30000:2000) is a diblock copolymer primarily utilized in the preparation of polymer micelles, vesicles, and other encapsulation applications. This product is intended for research use only.

  • Diblock copolymer
  • Used to prepare polymer micelles or vesicles
  • Suitable for encapsulation applications
  • Appearance: solid
  • Color: white to off-white
  • Purity: 90.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.