Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(styrene-divinylbenzene), 2% cross-linked, 30-60 mesh, Thermo Scientific™

Catalog No. AAA1756636
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1756636 500 g
AAA1756622 100 g
AAA175660E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1756636 Supplier Thermo Scientific Chemicals Supplier No. A1756636

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-70-7
Molecular Formula C24H27NO5
Molecular Weight (g/mol) 409.48
MDL Number 9003-70-7
InChI Key BCIPGSZQUDLGSY-UHFFFAOYNA-N
Synonym styrene/divinylbenzen, divinylbenzene-styrene, styrene divinylbenzene, styrene-divinylbenzene, styrene/divinylbenzene, styrene-divinyl benzene, benzene, diethenyl-, polymer with ethenylbenzene, brominated, st dvb, divinylbenzene; styrene, 1,2-diethenylbenzene; styrene
PubChem CID 174664
SMILES CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C=O

Specifications

Quantity 500 g
IUPAC Name tert-butyl 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoate
Mesh Size 30-60 Mesh
Odor Odorless
Chemical Name or Material Poly(styrene-divinylbenzene), 2% cross-linked
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.