Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(styrenesulfonic acid sodium salt) for for GPC 150,000, MilliporeSigma™ Supelco™

Catalog No. 111015885 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5885 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5885 Supplier MilliporeSigma Supelco Supplier No. 81614250MG
Add to Cart
Add to Cart

Specifications

CAS 9080-79-9
Physical Form Neat
MDL Number MFCD00084449
Synonym Poly(sodium styrenesulfonate)
Type Analytical Standard
Quantity 250 mg
Grade GPC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.