Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products GL401 Polyurethane Coated Nylon Gloves

Catalog No. 19555051 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-555-051 X-Large
19-555-049 Small
19-555-048 Medium
19-555-047 Large
19-555-050 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-555-051 Supplier Pyramex Safety Products Supplier No. GL401XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are designed to enhance grip capabilities without being sticky, in addition to providing a high degree of dexterity and tactile sensitivity.

  • 13 gauge nylon gloves with polyurethane coating
  • Provides dexterity and tactile sensitivity
  • Enhanced grip capabilities
  • Low particulate shed

Specifications

ANSI Abrasion Rating ANSI 1
Certifications/Compliance EN 388: 4131X, ANSI/ISEA 105-16 Level 2, ANSI/ISEA 105 A1
Certifications/Compliance (Europe) EN 388:4131X
Chemotherapeutic Rated No
Construction Seamless Knit
Cuff Material Latex Elastic
Coating Material Polyurethane
Color Gray
Exterior Finish Smooth
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size X-Large
EN388 Standard 4131X
For Use With (Application) Construction, Electronics, Inspection, and Quality Control
Gender Unisex
Glove Cuff Style Knit
Grip Design Smooth
Liner Material Nylon
Puncture Rating ANSI/ISEA 105-16 Level 2
Series GL401
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.