Learn More
Pyramex Safety Products GL402C5 Series Polyurethane Palm-Coated Gloves
Shop All Pyramex Safety Products ProductsDescription
- 13-gauge cut-resistant HPPE liner
- Low particulate shed
- Grips well without being sticky
- Offers tactile sensitivity and dexterity
- ANSI Cut Level: 4
- Abrasion Rating: 4
- Circular Blade Cut Rating: 5
- Tear Rating: 4
- Puncture Rating: 3
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 3 |
| Coating Material | Polyurethane |
| Color | Gray, Green |
| Cut Resistance Rating | ANSI A4 |
| Hand | Specific |
| Material | Polyurethane |
| Product Type | Cut-resistant Gloves |
| Size | X-Large |
| EN388 Standard | EN 388:4X43D |
| For Use With (Application) | Electronics, general duty, parts assembly, sheet metal, handling, glass cutting |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.