Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly-TPD

Catalog No. 9070651G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-706-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-706-51G Supplier Sigma Aldrich Supplier No. 9070651G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly-TPD is a solution processable polymer hole transport material. It's applications include:Organic Light Emitting Diode (OLED): Hole transport layer / Electron blocking layer (HTL/EBL) MaterialsSolution-Processed OLED : Polymer hole transport layerDye-Sensitized Solar Cell (DSSC):Hole transport materialsPerovskite Solar Cells:Hole transport material Flexible Printed Electronics:Hole transport layer / Hole injection layer(HTL/HIL) Polymers

Specifications

CAS 472960-35-3
Linear Formula (C22H21N)n
Quantity 1 g
Synonym 4-Butyl-N,N-diphenylaniline homopolymer; Poly(4-butyl-N,N-diphenylaniline); Poly(4-butyltriphenylamine); Poly[N,N-bis(4-butylphenyl) -N,N-bis(phenyl)-benzidine]
Molecular Weight (g/mol) 20000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.