Learn More
Polysciences, Inc. PolyTransport Buffer, 250 mL
SDP
Catalog No. NC1469091
Encompass_Preferred
Specially formulated transport medium designed to preserve tissue in a viable state during transport from surgery to grossing by maintaining osmolarity and pH for 24 to 48 hours.
Catalog No. NC1469091 Supplier Polysciences, Inc. Supplier No. 24311250
Specially formulated transport medium designed to preserve tissue in a viable state during transport from surgery to grossing by maintaining osmolarity and pH for 24 to 48 hours.