Learn More
INDEPENDENT INK INC POLY TREAT CHECK PENS
Catalog No. NC9914400
Encompass
This item has a minimum qty of 7 per supplier requirements.
Poly Treat Check Pen, If after five seconds the blue line remains bold and homogeneous, material has been correctly treated, If the treatment is borderline, The blue line will tend to bead up
Catalog No. NC9914400 Supplier INDEPENDENT INK INC Supplier No. PENS
This item has a minimum qty of 7 per supplier requirements.
Poly Treat Check Pen, If after five seconds the blue line remains bold and homogeneous, material has been correctly treated, If the treatment is borderline, The blue line will tend to bead up